Anti-Amitryptiline Mouse Monoclonal Antibody [clone: AM10-106.9]
ABNOMAB8238
: Abnova
- Antibody type:Primary
- Antigen name:Amitryptiline
- Clonality:Monoclonal
- Clone:AM10-106.9
- Conjugation:Unconjugated
- Isotype:IgG1
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS
- Storage temperature:Store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Amitriptyline conjugated with KLH.
- Pk:100 µG
Immunogen: Amitriptyline conjugated with KLH.
- For research use only (RUO)
Type: Primary
Antigen: Amitryptiline
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: