Anti-CD8A Rat Monoclonal Antibody (Spectral Red®) [clone: 53-6.7]
Catalog # ABNOMAB6109
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD8 antigen, alpha chain
- Antigen symbol:CD8A
- Clonality:Monoclonal
- Clone:53-6.7
- Conjugation:Spectral Red®
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:No
- Format:Liquid
- Gene ID:12525
- Antigen synonyms:p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
- Storage buffer:PBS (1 mg/mL sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified from mouse thymus/spleen cells.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Rat monoclonal antibody raised against native Cd8a.
Recommended Dilutions: Flow Cytometry: 0.2 ?g/10^6 cells
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: Spectral Red®
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse