Order Entry
Export
ContactUsLinkComponent
Anti-CD8A Rat Monoclonal Antibody (Cy5®) [clone: 53-6.7]
Anti-CD8A Rat Monoclonal Antibody (Cy5®) [clone: 53-6.7]
Catalog # ABNOMAB6110
Supplier:  Abnova
Anti-CD8A Rat Monoclonal Antibody (Cy5®) [clone: 53-6.7]
Catalog # ABNOMAB6110
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD8 antigen, alpha chain
  • Antigen symbol:
    CD8A
  • Clonality:
    Monoclonal
  • Clone:
    53-6.7
  • Conjugation:
    Cy5®
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    12525
  • Antigen synonyms:
    p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
  • Storage buffer:
    PBS (1 mg/mL sodium azide)
  • Storage temperature:
    Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified from mouse thymus/spleen cells.
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Rat monoclonal antibody raised against native Cd8a.

Recommended Dilutions: Flow Cytometry: 0.2 ?g/10^6 cells

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: Cy5®
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse