Anti-IL1R1 Mouse Monoclonal Antibody [clone: IL1-31-104.1.1]
ABNOMAB8354
: Abnova
- Antibody type:Primary
- Antigen name:Interleukin 1 Receptor
- Antigen symbol:IL1R1
- Clonality:Monoclonal
- Clone:IL1-31-104.1.1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:interleukin-1 receptor alpha|P80|antigen CD121a|IL-1R-alpha|IL-1RT1|IL-1R-1|IL1R|interleukin-1 receptor type 1|D2S1473|CD121 antigen-like family member A|interleukin-1 receptor type I|IL-1RT-1|interleukin 1 receptor alpha|CD121A|interleukin receptor 1|IL1RA|type I
- Storage buffer:PBS, pH 7,4
- Storage temperature:Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:The extracellular domain of the type I human IL-1 receptor (Genzyme Diagnostics).
- Tested applications:Radio Immunoassay
- Pk:200 µG
Mouse monoclonal antibody raised against partial IL-1 receptor.
Recommended Dilutions: ELISA: 5 ?g/ml Radio Immunoassay: 20-30 ng/ml; Immunohistochemistry: 10-100 ?g/ml
Type: Primary
Antigen: IL1R1
Clonality: Monoclonal
Clone: IL1-31-104.1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human