Anti-Insulin Receptor Monoclonal Antibody [clone: 47-9]
ABNOMAB8375
: Abnova
- Antibody type:Primary
- Antigen name:Insulin Receptor
- Clonality:Monoclonal
- Clone:47-9
- Conjugation:Unconjugated
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7.0
- Storage temperature:Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified from IM-9 lymphocytes.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against native Insulin receptor.
Type: Primary
Antigen: Insulin Receptor
Clonality: Monoclonal
Clone: 47-9
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: Human