Order Entry
Export
ContactUsLinkComponent
Anti-BGLAP Mouse Monoclonal Antibody [clone: OC4-30]
Anti-BGLAP Mouse Monoclonal Antibody [clone: OC4-30]
Catalog # ABNOMAB8376
Supplier:  Abnova
Anti-BGLAP Mouse Monoclonal Antibody [clone: OC4-30]
Catalog # ABNOMAB8376
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Bone Gamma-Carboxyglutamate Protein
  • Antigen symbol:
    BGLAP
  • Clonality:
    Monoclonal
  • Clone:
    OC4-30
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Bovine
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Lyophilized
  • Gene ID:
    281646
  • Antigen synonyms:
    BGP
  • Storage buffer:
    10 mM PBS, pH 7,4 (1% BSA, 0,1% sodium azide)
  • Storage temperature:
    Store at 4 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Bovine BGLAP.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against BGLAP.

Recommended Dilutions: Sandwich ELISA: 1-10 ?g/ml; Western Blot: 1-10 ?g/ml; Immunohistochemistry (Frozen sections): 1-10 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1-10 ?g/ml

Type: Primary
Antigen: BGLAP
Clonality: Monoclonal
Clone: OC4-30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine