Anti-Monocyte/Granulocyte Mouse Monoclonal Antibody [clone: 74-22-15]
Catalog # ABNOMAB6219
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Monocyte/Granulocyte
- Antigen symbol:Monocyte/Granulocyte
- Clonality:Monoclonal
- Clone:74-22-15
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Pig
- Cross adsorption:No
- Format:Liquid
- Storage buffer:100 mM BBS, pH 8,2
- Storage temperature:Store at 4 °C.
- Immunogen:dd Miniature swine thymocytes.
- Size:500 μg
- Pk:500 µg
Specifications
About this item
Mouse monoclonal antibody raised against Monocyte/Granulocyte.
Type: Primary
Antigen: Monocyte/Granulocyte
Clonality: Monoclonal
Clone: 74-22-15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pig