Anti-biogenic amine (5HT) Receptor Rabbit Polyclonal Antibody
Catalog # ABNOPAB16035
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:biogenic amine (5HT) Receptor
- Antigen symbol:biogenic amine (5HT) Receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,2 (50% glycerol, 0,01% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to N-terminus of Schistosoma mansoni biogenic amine (5HT) receptor.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of biogenic amine (5HT) receptor.
Recommended Dilutions: ELISA: 1:2000-1:5000; Western Blot: 1 ?g/ml
Type: Primary
Antigen: biogenic amine (5HT) Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: