Anti-RIF1 Goat Polyclonal Antibody
Catalog # ABNOPAB6700
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:RAP1 interacting factor
- Antigen symbol:RIF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Cross adsorption:No
- Format:Liquid
- Gene ID:55183
- Antigen synonyms:DKFZp781N1478|rap1-interacting factor 1 homolog|RAP1 interacting factor homolog|FLJ12870|yeast|telomere-associated protein RIF1|DKFZp434D1026
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:CEEGIIDANKTETNT
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human RIF1.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of RIF1.
Recommended Dilutions: ELISA: 1:16000
Type: Primary
Antigen: RIF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: