Anti-EPHA2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB18348
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ephrin type A receptor 2
- Antigen symbol:EPHA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:1969|2042|2043
- Antigen synonyms:Epithelial cell kinase|CTPA|ARCC2|ECK|CTRCT6|CTPP1|Ephrin type-A receptor 2|Tyrosine-protein kinase receptor ECK
- Storage buffer:PBS, 150mM NaCl, pH 7,4 (50% glycerol, 0,02% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to residues surrounding Y588/Y596 of human EPHA2/EPHA3/EPHA4.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of EPHA2/EPHA3/EPHA4.
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunofluorescence: 1:500-1:1000; ELISA: 1:40000
Type: Primary
Antigen: EPHA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat