Anti-KIR2DL4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: mAb 33]
Catalog # ABNOMAB9848
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Killer cell immunoglobulin-like Receptor two Domains long cytoplasmic tail 4
- Antigen symbol:KIR2DL4
- Clonality:Monoclonal
- Clone:mAb 33
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:3805
- Antigen synonyms:KIR103|killer cell immunoglobulin-like receptor 2DL4|natural killer cell inhibitory receptor|killer Ig receptor|killer cell inhibitory receptor 103AS|MHC class I NK cell receptor KIR103AS|CD158 antigen-like family member D|NK cell receptor|KIR103AS|CD158D|G9P|KIR-103AS
- Storage buffer:PBS, pH 7,4 (0,02% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Avoid prolonged exposure to light. Do not freeze.
- Immunogen:NK3.3 cells and KIR2DL4-Ig fusion protein.
- Purification:Size-exclusion chromatography purification
- Size:100 Reactions
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against KIR2DL4.
Recommended Dilutions: Flow Cytometry: 20 ?l reagent/100 ?l of whole blood or 10^6 cells
Type: Primary
Antigen: KIR2DL4
Clonality: Monoclonal
Clone: mAb 33
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human