Anti-ANXA1 Mouse Monoclonal Antibody [clone: 39]
Catalog # ABNOMAB9893
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Annexin A1
- Antigen symbol:ANXA1
- Clonality:Monoclonal
- Clone:39
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:301
- Antigen synonyms:ANX1|LPC1
- Storage buffer:Citrate-Tris-HCl buffer, pH 7.0 (0,02% Procl300)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human ANXA1.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant ANXA1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: ANXA1
Clonality: Monoclonal
Clone: 39
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human