Order Entry
Export
ContactUsLinkComponent
Anti-ERP29 Mouse Monoclonal Antibody [clone: 39]
Anti-ERP29 Mouse Monoclonal Antibody [clone: 39]
Catalog # ABNOMAB9894
Supplier:  Abnova
Anti-ERP29 Mouse Monoclonal Antibody [clone: 39]
Catalog # ABNOMAB9894
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Endoplasmic Reticulum Protein 29
  • Antigen symbol:
    ERP29
  • Clonality:
    Monoclonal
  • Clone:
    39
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    10961
  • Antigen synonyms:
    HEL-S-107|ERp31|ERp28|C12orf8|PDIA9|PDI-DB
  • Amino acid number:
    33 to 261
  • Storage buffer:
    Citrate-Tris-HCl buffer, pH 7.0 (0,02% Procl300)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids 33-261 of human ERP29.
  • Purification:
    Affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant ERP29.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ERP29
Clonality: Monoclonal
Clone: 39
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human