Anti-TUBB Mouse Monoclonal Antibody [clone: 37]
Catalog # ABNOMAB9930
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Tubulin beta
- Antigen symbol:TUBB
- Clonality:Monoclonal
- Clone:37
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:203068
- Antigen synonyms:tubulin beta polypeptide|tubulin beta-5 chain|M40|beta Ib tubulin|tubulin|beta 5-tubulin|tubulin beta chain|tubulin beta-1 chain|CDCBM6|beta polypeptide|beta-4 tubulin|OK/SW-cl.56|TUBB1|TUBB5
- Amino acid number:400 to 444
- Storage buffer:Citrate-Tris-HCl buffer, pH 7.0 (0,02% Procl300)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 400-444 of human TUBB.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant TUBB.
Type: Primary
Antigen: TUBB
Clonality: Monoclonal
Clone: 37
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human