Anti-PEBP1 Mouse Monoclonal Antibody [clone: 21]
ABNOMAB9940
: Abnova
- Antibody type:Primary
- Antigen name:Phosphatidylethanolamine Binding Protein 1
- Antigen symbol:PEBP1
- Clonality:Monoclonal
- Clone:21
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:5037
- Antigen synonyms:HCNPpp|PEBP|HEL-S-34|PBP|RKIP|PEBP-1|HCNP|HEL-210
- Storage buffer:Citrate-Tris-HCl buffer, pH 7.0 (0,02% Procl300)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human PEBP1.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against full length recombinant PEBP1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: PEBP1
Clonality: Monoclonal
Clone: 21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human