Order Entry
Export
ContactUsLinkComponent
 
Anti-PRDX6 Mouse Monoclonal Antibody [clone: 52]
  ABNOMAB9948
 :  Abnova
Anti-PRDX6 Mouse Monoclonal Antibody [clone: 52]
  ABNOMAB9948
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Peroxiredoxin 6
  • Antigen symbol:
    PRDX6
  • Clonality:
    Monoclonal
  • Clone:
    52
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    9588
  • Antigen synonyms:
    KIAA0106|MGC46173|1-Cys PRX|AOP2|NSGPx|liver 2D page spot 40|peroxiredoxin-6|p29|antioxidant protein 2|1-Cys|PRX|non-selenium glutathione peroxidase|24 kDa protein|red blood cells page spot 12|aiPLA2|1-Cys peroxiredoxin|acidic calcium-independent phospholipase A2
  • Storage buffer:
    Citrate-Tris-HCl buffer, pH 7.0 (0,02% Procl300)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to full length human PRDX6.
  • Purification:
    Affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against full length recombinant PRDX6.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PRDX6
Clonality: Monoclonal
Clone: 52
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human