Anti-CLU Mouse Monoclonal Antibody [clone: 16]
Catalog # ABNOMAB9962
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:clusterin
- Antigen symbol:CLU
- Clonality:Monoclonal
- Clone:16
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:1191
- Antigen synonyms:SGP-2|NA1/NA2|KUB1|SP-40|TRPM-2|SGP2|CLU1|APOJ|CLI|APO-J|TRPM2|AAG4|CLU2
- Amino acid number:23 to 449
- Storage buffer:Citrate-Tris-HCl buffer, pH 7.0 (0,02% Procl300)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 23-449 of human CLU.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant CLU.
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: CLU
Clonality: Monoclonal
Clone: 16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human