Anti-CAPZA2 Mouse Monoclonal Antibody [clone: 40]
Catalog # ABNOMAB9969
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Capping protein (actin filament) muscle Z-line, alpha 2
- Antigen symbol:CAPZA2
- Clonality:Monoclonal
- Clone:40
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:830
- Antigen synonyms:CAPPA2|alpha 2|CapZ alpha 2|CAZA2_HUMAN.|F actin capping protein subunit alpha 2|Capping protein actin filament muscle Z line|CAPZ
- Storage buffer:Citrate-Tris-HCl buffer, pH 7.0 (0,02% Procl300)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human CAPZA2.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 Tests
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant CAPZA2.
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: CAPZA2
Clonality: Monoclonal
Clone: 40
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human