Order Entry
Export
ContactUsLinkComponent
 
Anti-FASLG Rabbit Polyclonal Antibody
  ABNOPAB18864
 :  Abnova
Anti-FASLG Rabbit Polyclonal Antibody
  ABNOPAB18864
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Fas Ligand
  • Antigen symbol:
    FASLG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    356
  • Antigen synonyms:
    APT1LG1|APTL|TNFSF6|CD178|FASL|ALPS1B|CD95-L|CD95L
  • Storage buffer:
    buffer containing 0,02% sodium azide
  • Storage temperature:
    Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to 15 amino acids of human FASLG.
  • Pk:
    200 µl

 

 

Rabbit polyclonal antibody raised against synthetic peptide of FASLG.

Recommended Dilutions: ELISA: 1:10000-1:40000; Western Blot: 1:100-1:500; Immunohistochemistry: 1:100-1:500

Type: Primary
Antigen: FASLG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human