Anti-AKR1B1 Mouse Monoclonal Antibody [clone: 19]
Catalog # ABNOMAB9983
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Aldo-keto reductase family 1 member B1
- Antigen symbol:AKR1B1
- Clonality:Monoclonal
- Clone:19
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:231
- Antigen synonyms:AKR1B1|ALR2|ADR|Aldo-keto reductase family 1 member B1|Aldose reductase|AR|Aldehyde reductase|ALDR1
- Amino acid number:33 to 316
- Storage buffer:Citrate-Tris-HCl buffer, pH 7.0 (0,02% Procl300)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 33-316 of human AKR1B1.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant AKR1B1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: AKR1B1
Clonality: Monoclonal
Clone: 19
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human