Anti-TMEM184B Rabbit Polyclonal Antibody
Catalog # ABNOPAB19229
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Transmembrane Protein 184B
- Antigen symbol:TMEM184B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:25829
- Antigen synonyms:Transmembrane protein 184B|Putative MAPK-activating protein FM08|PSEC0108|C22orf5|TMEM184B|HS5O6A|FM08
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 15 amino acids near C-terminus of human TMEM184B.
- Purification:Peptide affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody rasied against synthetic peptide of TMEM184B.
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry: 5 ?g/ml
Type: Primary
Antigen: TMEM184B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat