Anti-SIRT1 Goat Polyclonal Antibody
Catalog # ABNOPAB19664
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Sirtuin 1
- Antigen symbol:SIRT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Epitope:Center
- Cross adsorption:No
- Format:Liquid
- Gene ID:23411
- Antigen synonyms:NAD-dependent Deacetylase Sirtuin-1|SIR2L1|SIRT1|SIR2-like Protein 1
- Amino acid number:577 to 590
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-EKPQEVQTSRNVES
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 577-590 at internal region of human SIRT1.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of SIRT1.
Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.05-0.2 ?g/ml
Type: Primary
Antigen: SIRT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity: Human