Anti-SLC46A1 Goat Polyclonal Antibody
Catalog # ABNOPAB19698
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 46 Member A1
- Antigen symbol:SLC46A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Epitope:Center
- Cross adsorption:No
- Format:Liquid
- Gene ID:113235
- Antigen synonyms:PCFT|HCP1|G21
- Amino acid number:233 to 247
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-LKEPKSTRLFTFRH
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 233-247 at internal region of human SLC46A1.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 Tests
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of SLC46A1.
Recommended Dilutions: Western Blot: 0.3-1 ?g/ml
Type: Primary
Antigen: SLC46A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity: Human