Order Entry
Export
ContactUsLinkComponent
Anti-17-alpha-OH-Progesterone Polyclonal Antibody
Anti-17-alpha-OH-Progesterone Polyclonal Antibody
Catalog # ABNOPAB8201
Supplier:  Abnova
Anti-17-alpha-OH-Progesterone Polyclonal Antibody
Catalog # ABNOPAB8201
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    17-alpha-OH-Progesterone
  • Antigen symbol:
    17-alpha-OH-Progesterone
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    20 mM phosphate buffer, 150 mM NaCl, pH 7,2 (0,09% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    17 alpha-OH-Progesterone-BTG.
  • Pk:
    1 mg

Specifications

About this item

Sheep polyclonal antibody raised against 17 alpha-OH-Progesterone.

Recommended Dilutions: ELISA: 0.31 ?g/ml

Type: Primary
Antigen: 17-alpha-OH-Progesterone
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: