Anti-GFRA2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB9888
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:GDNF family receptor alpha 2
- Antigen symbol:GFRA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:2675
- Antigen synonyms:GDNF family receptor alpha-2|NTNRA|RETL2|GDNFRB|GDNF receptor alpha-2|GDNF receptor beta|TRNR2|GFR alpha 2|NRTNR-ALPHA
- Amino acid number:377 to 391
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 377-391 of human GFRA2.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of GFRA2.
Recommended Dilutions: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: GFRA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat