Order Entry
Export
ContactUsLinkComponent
Anti-CCNB1 Rabbit Polyclonal Antibody
Anti-CCNB1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB9943
Supplier:  Abnova
Anti-CCNB1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB9943
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclin B1
  • Antigen symbol:
    CCNB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Antiserum
  • Gene ID:
    891
  • Antigen synonyms:
    CCNB
  • Storage buffer:
    0,01% sodium azide
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant GST fusion protein corresponding to full length human CCNB1.
  • Size:
    100 μl
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against full length recombinant CCNB1.

Recommended Dilutions: ELISA: 1:2000-1:10000; Western Blot: 1:500-1:1000

Type: Primary
Antigen: CCNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat