Anti-GHSR Rabbit Polyclonal Antibody
50 μg
Catalog # ABNOPAB26268
Supplier: Abnova
Jump to:
Product Details & Documents
Rabbit polyclonal antibody raised against synthetic peptide of GHSR.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 5-7.5 ?g/ml
Type: Primary
Antigen: GHSR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gorilla, Human
Documents
TDS_ABNOPAB26268.pdf
Datasheet
Specifications
- Size:50 μg
- Clonality:Polyclonal
- Purification:Immunoaffinity chromatography
- Antibody type:Primary
- Storage temperature:Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Host:Rabbit
- Cross adsorption:No
- Antigen name:Growth hormone secretagogue receptor
- Cat. No.:ABNOPAB26268
- Reactivity:Gorilla|Human
- Format:Liquid
- Gene ID:2693
- Antigen synonyms:GHSR|growth hormone secretagogue receptor|GHSR1|GHDP
- Immunogen:A synthetic peptide corresponding to 13 amino acid at N-terminus of human GHSR.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:GHSR
- Storage buffer:PBS (0,1% sodium azide)
- ELISA:Yes



