Order Entry
Export
ContactUsLinkComponent
Anti-ADGRE1 Rabbit Polyclonal Antibody
Anti-ADGRE1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB26248
Supplier:  Abnova
Anti-ADGRE1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB26248
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Adhesion G Protein-Coupled Receptor E1
  • Antigen symbol:
    ADGRE1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    2015
  • Antigen synonyms:
    Ly71|Gpf480|F4/80|TM7LN3|DD7A5-7|EGF-TM7|Emr1
  • Storage buffer:
    PBS (0,1% sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to 19 amino acid at N-terminus of human EMR1.
  • Purification:
    Immunoaffinity chromatography
  • Size:
    50 μg
  • Pk:
    50 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of EMR1.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 18-36 ?g/ml

Type: Primary
Antigen: ADGRE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human