Order Entry
Export
ContactUsLinkComponent
Anti-MTOR Rabbit Polyclonal Antibody
Anti-MTOR Rabbit Polyclonal Antibody
Catalog # ABNOPAB25827
Supplier:  Abnova
Anti-MTOR Rabbit Polyclonal Antibody
Catalog # ABNOPAB25827
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mechanistic Target of Rapamycin
  • Antigen symbol:
    MTOR
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    2475
  • Antigen synonyms:
    FRAP1|mTOR|m-TOR|RAPT1|RAFT1|FKBP-Rapamycin Associated Protein|Mechanistic Target Of Rapamycin|EC 2.7.11.1|FRAP|FRAP2|Rapamycin Target Protein|FK506 binding protein 12-rapamycin associated protein|Mammalian Target Of Rapamycin|Serine/Threonine-Protein Kinase MTOR|MTOR
  • Modification:
    phospho-Ser2448
  • Storage buffer:
    PBS (without Mg(2+) and Ca(2+)), 150 mM NaCl, pH 7,4 (50% glycerol, 0,02% sodium azide)
  • Sequence:
    T-D-Sp-Y-S
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Synthetic phosphopeptide corresponding to residues surrounding S2448 of human MTOR.
  • Purification:
    purified by Affinity Chromatography
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic phosphopeptide of MTOR.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: MTOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat