Anti-CFL1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB25817
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cofilin 1
- Antigen symbol:CFL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:1072
- Antigen synonyms:HEL-S-15|cofilin|CFL
- Modification:phospho-Ser3
- Storage buffer:PBS (without Mg(2+) and Ca(2+)), 150 mM NaCl, pH 7,4 (50% glycerol, 0,02% sodium azide)
- Sequence:M-A-Sp-G-V
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding S3 of human CFL1.
- Purification:purified by Affinity Chromatography
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CFL1.
Recommended Dilutions: Immunohistochemistry: 1:50-1:100; Western Blot: 1:500-1:1000
Type: Primary
Antigen: CFL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat