Order Entry
Export
ContactUsLinkComponent
 
Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7) [clone: OKT3]
 
Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7) [clone: OKT3]

 

  • Catalog No:
  • 60-0037-T025
  • 60-0037-T100
  • Antigen symbol:
  • CD3E
  • CD3E
  • Antigen name:
  • CD3 Molecule Epsilon
  • CD3 Molecule Epsilon
  • Antigen synonyms:
  • CD3e antigen|TCRE|epsilon subunit of T3|epsilon polypeptide TiT3 complex|T3E|FLJ18683|CD3-epsilon|T-cell surface glycoprotein CD3 epsilon chain|T-cell antigen receptor complex|T-cell surface antigen T3/Leu-4 epsilon chain
  • CD3e antigen|TCRE|epsilon subunit of T3|epsilon polypeptide TiT3 complex|T3E|FLJ18683|CD3-epsilon|T-cell surface glycoprotein CD3 epsilon chain|T-cell antigen receptor complex|T-cell surface antigen T3/Leu-4 epsilon chain
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)-Cy7®
  • PE (Phycoerythrin)-Cy7®
  • Clone:
  • OKT3
  • OKT3
  • Reactivity:
  • Chimpanzee, Human
  • Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 916
  • 916
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • 2...8 °C, protected from light
  • 2...8 °C, protected from light
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry