Anti-CD2 Rat Monoclonal Antibody (In Vivo Ready™) [clone: RM2-5]
40-0021-U100
- Antibody type:Primary
- Antigen name:CD2 molecule
- Antigen symbol:CD2
- Clonality:Monoclonal
- Clone:RM2-5
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b lambda
- Reactivity:Mouse
- Gene ID:12481
- Antigen synonyms:sheep red blood cell receptor|T-cell surface antigen T11|erythrocyte receptor|rosette receptor|T11|Leu-5|SRBC|LFA-3 receptor|lymphocyte-function antigen-2|LFA-2
- Storage temperature:2...8 °C
- Tested applications:Functional Assay
- Size:100 µg
- Pk:100 µG
The RM2-5 antibody reacts with mouse CD2, an approximately 50 kDa glycoprotein, and a member of the Ig superfamily. CD2, also known as LFA-2, is a receptor for CD48 in the mouse and is expressed on the cell surface of all lymphocytes. CD2 mediates cell to cell adhesion and is involved in T cell activation.
RM2-5 is reported to block CD2-mediated cell:cell adhesion. Please choose the appropriate format for each application.
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RM2-5
Conjugation: In Vivo Ready™
Epitope:
Host: Rat
Isotype: IgG2b, lambda
Reactivity: Mouse