About this item
The IM7 antibody recognizes CD44, a ubiquitously expressed cell surface receptor which is important for extracellular matrix organization, cell-cell and cell-matrix adhesion and migration. CD44 may be expressed in a number of different isoforms (splice variants) from the most typical or “standard” form, known as CD44s, to variants designated CD44v, e.g. CD44v1 or CD44v6. These receptors interact with several ligands, but most often associate with an extracellular matrix component hyaluronate, through which it mediates adhesion.
The IM7 antibody may be used for detection of all isoforms of CD44, as it recognizes constant epitopes near the extracellular proximal domain. (Xu et al, 2002, J. Leukoc. Biol. 72:1133-1141). It has been reported to be cross-reactive with many non-human species including Baboon, Chimpanzee, Cynomolgus, Rhesus, Horse, Cow, Pig, Dog and Cat CD44.
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: redFluor™ 710
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Human, Mouse, Chimpanzee, Baboon, Cynomolgus, Rhesus, Pig, Cat, Dog, Horse
Specifications
- Catalog No:
- 80-0441-U025
- 80-0441-U100
- Antigen symbol:
- CD44
- CD44
- Antigen name:
- CD44 antigen
- CD44 antigen
- Antigen synonyms:
- HERMES|AW121933|AW146109|AU023126|Pgp-1|Ly-24
- HERMES|AW121933|AW146109|AU023126|Pgp-1|Ly-24
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- redFluor® 710
- redFluor® 710
- Clone:
- IM7
- IM7
- Reactivity:
- Baboon, Pig, Horse, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Dog, Mouse, Cat
- Baboon, Pig, Horse, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Dog, Mouse, Cat
- Host:
- Rat
- Rat
- Gene ID:
- 12505
- 12505
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- 2...8 °C, protected from light
- 2...8 °C, protected from light
- Tested applications:
- Flow Cytometry
- Flow Cytometry