Order Entry
Export
ContactUsLinkComponent
Anti-CD45 Rat Antibody (redFluor™ 710) [clone: 30-F11]
Anti-CD45 Rat Antibody (redFluor™ 710) [clone: 30-F11]
Supplier:  Tonbo Biosciences
Certificates
Anti-CD45 Rat Antibody (redFluor™ 710) [clone: 30-F11]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 80-0451-U100
  • 80-0451-U025
  • Antigen symbol:
  • PTPRC
  • PTPRC
  • Antigen name:
  • Protein Tyrosine Phosphatase Receptor Type C
  • Protein Tyrosine Phosphatase Receptor Type C
  • Antigen synonyms:
  • CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • Antibody type:
  • Primary
  • Primary
  • Conjugation:
  • redFluor® 710
  • redFluor® 710
  • Clone:
  • 30-F11
  • 30-F11
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 19264
  • 19264
  • Isotype:
  • IgG2b
  • IgG2b
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage temperature:
  • Store undiluted at 2...8 °C; Protect from Light; Do not freeze
  • Store undiluted at 2...8 °C; Protect from Light; Do not freeze
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information