Order Entry
Export
Orders LinkContactUsLinkComponent
Anti-GPATCH11 Rabbit Polyclonal Antibody

Anti-GPATCH11 Rabbit Polyclonal Antibody

Catalog # ABGEAP18182A
Supplier:   Abgent
Anti-GPATCH11 Rabbit Polyclonal Antibody
Catalog # ABGEAP18182A
Supplier:  Abgent
Anti-GPATCH11 Rabbit Polyclonal Antibody
Price
365,00 €
Log in for availability
1 * 400 µl
Product Details & Documents

Type: Primary
Antigen: GPATCH11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Hamster, Mouse

Documents

Specifications
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Protein A & Peptide affinity purification
  • Antibody type:
    Primary
  • Epitope:
    N-terminal
  • Amino acid number:
    12 to 41
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Host:
    Rabbit
  • Sequence:
    RQIREARRKEEKQQEC
  • Concentration:
    1.280
  • Molecular weight:
    30.199 kDa
  • Shipping temperature:
    4 °C
  • Antigen name:
    G Patch Domain Containing 11
  • Cat. No.:
    ABGEAP18182A
  • Reactivity:
    Hamster|Human|Mouse
  • Gene ID:
    253635
  • Antigen synonyms:
    CENP-Y|CENPY|CCDC75
  • Immunogen:
    This CCDC75 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 12-41 amino acids from the N-terminal region of human CCDC75.
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    GPATCH11
  • Storage buffer:
    pH 7,4
  • ELISA:
    Yes