Anti-Sh2d5 Rabbit Polyclonal Antibody
Catalog # ABGEAP20512C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen symbol:Sh2d5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Mouse
- Western blot:Yes
- Epitope:internal
- Amino acid number:200 to 230
- Storage buffer:pH 7,4
- Molecular weight:38.174 kDa
- Sequence:PESEGRGGTRHIRC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5
- Shipping temperature:4 °C
- Immunogen:This Mouse Sh2d5 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 200-230 amino acids from the Central region of mouse Sh2d5.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: Sh2d5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Mouse