Anti-ZNF695 Rabbit Polyclonal Antibody
Catalog # ABGEAP17947C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:zinc finger protein 695
- Antigen symbol:ZNF695
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Gene ID:57116
- Antigen synonyms:SBZF3
- Amino acid number:244 to 269
- Storage buffer:pH 7,4
- Molecular weight:60.149 kDa
- Sequence:EKNHTEKKTYRC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.6
- Shipping temperature:4 °C
- Immunogen:This ZNF695 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 244-269 amino acids from the Central region of human ZNF695.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: ZNF695
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human