Anti-MYH14 Rabbit Polyclonal Antibody
Catalog # ABGEAP20565A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:myosin, heavy chain 14, non-muscle
- Antigen symbol:MYH14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Gene ID:79784
- Antigen synonyms:FP17425|DFNA4A|NMHC-II-C|MYH17|MHC16|PNMHH|DFNA4|NMHC II-C|myosin
- Storage buffer:pH 7,4
- Molecular weight:227.871 kDa
- Sequence:GDGPPGGRPRRGMFR
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.49
- Shipping temperature:4 °C
- Immunogen:This MYH14 antibody is generated from a rabbit immunized with a KLH conjugated synthetic peptide between 654-668 amino acids from the Central region of human MYH14.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: MYH14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human