Order Entry
Export
ContactUsLinkComponent
Anti-HSDL2 Rabbit Polyclonal Antibody
Anti-HSDL2 Rabbit Polyclonal Antibody
  ABGEAP16340C
 :  Abgent
Anti-HSDL2 Rabbit Polyclonal Antibody
  ABGEAP16340C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    hydroxysteroid dehydrogenase like 2
  • Antigen symbol:
    HSDL2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    internal
  • Gene ID:
    84263
  • Antigen synonyms:
    C9orf99|SDR13C1
  • Amino acid number:
    229 to 258
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    45.395 kDa
  • Sequence:
    IDENILKEEGIENFDC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.65
  • Shipping temperature:
    4 °C
  • Immunogen:
    This HSDL2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 229-258 amino acids from the Central region of human HSDL2.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: HSDL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human