Anti-PPP1CB Rabbit Polyclonal Antibody
Catalog # ABGEAP20422B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen symbol:PPP1CB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Gene ID:443852
- Amino acid number:276 to 304
- Storage buffer:pH 7,4
- Molecular weight:37.173 kDa
- Sequence:SFQILKPSEKKAKYC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.62
- Shipping temperature:4 °C
- Immunogen:This XENLA ppp1cb antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 276-304 amino acids from the C-terminal region of human XENLA ppp1cb.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: PPP1CB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human