Anti-RNF219 Rabbit Polyclonal Antibody
Catalog # ABGEAP17601C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Ring Finger Protein 219
- Antigen symbol:RNF219
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Gene ID:79596
- Antigen synonyms:C13orf7
- Amino acid number:275 to 302
- Storage buffer:pH 7,4
- Molecular weight:81.116 kDa
- Sequence:VSKNQGDSARKQPC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.36
- Shipping temperature:4 °C
- Immunogen:This RNF219 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 275-302 amino acids from the Central region of human RNF219.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: RNF219
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human