Anti-ANKDD1A Rabbit Polyclonal Antibody
Catalog # ABGEAP19734B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:ankyrin repeat and death domain containing 1A
- Antigen symbol:ANKDD1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Gene ID:348094
- Amino acid number:382 to 410
- Storage buffer:pH 7,4
- Molecular weight:57.55 kDa
- Sequence:IGRRDLAENIRKKAC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.64
- Shipping temperature:4 °C
- Immunogen:This ANKDD1A antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 382-410 amino acids from the C-terminal region of human ANKDD1A.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: ANKDD1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human