Order Entry
Export
ContactUsLinkComponent
Anti-TP53BP2 Rabbit Polyclonal Antibody
Anti-TP53BP2 Rabbit Polyclonal Antibody
Catalog # ABGEAP16353A
Supplier:  Abgent
Anti-TP53BP2 Rabbit Polyclonal Antibody
Catalog # ABGEAP16353A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tumor Protein p53 Binding Protein 2
  • Antigen symbol:
    TP53BP2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Gene ID:
    7159
  • Antigen synonyms:
    P53BP2|ASPP2|PPP1R13A|53BP2|BBP
  • Amino acid number:
    93 to 122
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    125.616 kDa
  • Sequence:
    VPGEYRRKENGVNSPC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.98
  • Shipping temperature:
    4 °C
  • Immunogen:
    This TP53BP2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 93-122 amino acids from the N-terminal region of human TP53BP2.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: TP53BP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human