Anti-TGFB2 Rabbit Polyclonal Antibody
Catalog # ABGEAP1476B
Supplier: Abgent
Jump to:
Product Details & Documents
Type: Primary
Antigen: TGFB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Documents
Specifications
- Western blot:Yes
- Clonality:Polyclonal
- Flow cytometry:Yes
- Purification:Saturated Ammonium Sulfate (SAS)
- Antibody type:Primary
- Epitope:C-terminal
- Amino acid number:277 to 306
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Host:Rabbit
- Sequence:CESQQTNRRKKRALDA
- Concentration:5.710
- Molecular weight:47.748 kDa
- Shipping temperature:4 °C
- Antigen name:transforming growth factor, beta 2
- Cat. No.:ABGEAP1476B
- Reactivity:Human
- Gene ID:7042
- Antigen synonyms:LDS4|TGF-beta2
- Immunogen:This TGF Beta 2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 277-306 amino acids from the C-terminal region of human TGF Beta 2.
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:TGFB2
- Storage buffer:pH 7,4
- ELISA:Yes



