Order Entry
Export
ContactUsLinkComponent
Anti-SNAP25 Rabbit Polyclonal Antibody
Anti-SNAP25 Rabbit Polyclonal Antibody
  ABGEAP15001A
 :  Abgent
Anti-SNAP25 Rabbit Polyclonal Antibody
  ABGEAP15001A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Synaptosomal-Associated Protein, 25kDa
  • Antigen symbol:
    SNAP25
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    6616
  • Antigen synonyms:
    SEC9|bA416N4.2|dJ1068F16.2|SNAP|SNAP-25|RIC-4|RIC4|CMS18
  • Modification:
    phospho-Thr138
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    23.315 kDa
  • Sequence:
    FIRRVUNDAREC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.49
  • Shipping temperature:
    4 °C
  • Immunogen:
    This SNAP25 Antibody is generated from rabbits immunized with a KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding T138 of human SNAP25.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: SNAP25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human