Anti-STAR Rabbit Polyclonal Antibody
ABGEAP16547C
: Abgent
- Antibody type:Primary
- Antigen name:Steroidogenic Acute Regulatory Protein
- Antigen symbol:STAR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Gene ID:6770
- Antigen synonyms:STARD1
- Amino acid number:88 to 117
- Storage buffer:pH 7,4
- Molecular weight:31.914 kDa
- Sequence:DNGDKVMSKVVPDVGC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.6
- Shipping temperature:4 °C
- Immunogen:This STAR antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 88-117 amino acids from the Central region of human STAR.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: STAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human