Order Entry
Export
ContactUsLinkComponent
Anti-ZNF865 Rabbit Polyclonal Antibody
Anti-ZNF865 Rabbit Polyclonal Antibody
Catalog # ABGEAP20132B
Supplier:  Abgent
Anti-ZNF865 Rabbit Polyclonal Antibody
Catalog # ABGEAP20132B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    zinc finger protein 865
  • Antigen symbol:
    ZNF865
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Gene ID:
    100507290
  • Amino acid number:
    1015 to 1042
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    111.077 kDa
  • Sequence:
    YGLKKHRLAHKAEC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.49
  • Shipping temperature:
    4 °C
  • Immunogen:
    This ZNF865 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1015-1042 amino acids from the C-terminal region of human ZNF865.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: ZNF865
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Mouse