Order Entry
Export
ContactUsLinkComponent
Anti-F13B Rabbit Polyclonal Antibody
Anti-F13B Rabbit Polyclonal Antibody
Catalog # ABGEAP20162A
Supplier:  Abgent
Anti-F13B Rabbit Polyclonal Antibody
Catalog # ABGEAP20162A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    coagulation factor XIII, B polypeptide
  • Antigen symbol:
    F13B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Gene ID:
    2165
  • Antigen synonyms:
    FXIIIB
  • Amino acid number:
    151 to 179
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    75.511 kDa
  • Sequence:
    TQKTFKVKDKVQYEC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.73
  • Shipping temperature:
    4 °C
  • Immunogen:
    This F13B antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 151-179 amino acids from the N-terminal region of human F13B.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: F13B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Mouse