Order Entry
Export
ContactUsLinkComponent
Anti-CRYAB Mouse Monoclonal Antibody [clone: 1329CT523.140.120]
Anti-CRYAB Mouse Monoclonal Antibody [clone: 1329CT523.140.120]
Catalog # ABGEAM8424B
Supplier:  Abgent
undefined
Anti-CRYAB Mouse Monoclonal Antibody [clone: 1329CT523.140.120]
Catalog # ABGEAM8424B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Crystallin, alpha B
  • Antigen symbol:
    CRYAB
  • Clonality:
    Monoclonal
  • Clone:
    1329CT523.140.120
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    1410
  • Antigen synonyms:
    HSPB5|CRYA2|CMD1II|CTPP2|HEL-S-101|CTRCT16|MFM2
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    20.159 kDa
  • Sequence:
    MDIAIHHPWIRRPFFPFHSPSRLFDQFFGEHLLESDLFPTSTSLSPFYLRPPSFLRAPSWFDTGLSEMRLEKDRFSVNLDVKHFSPEELKVKVLGDVIEVHGKHEERQDEHGFISREFHRKYRIPADVDPLTITSSLSSDGVLTVNGPRKQVSGPERTIPITREEKPAVTAAPKK
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    1.230
  • Shipping temperature:
    4 °C
  • Immunogen:
    This CRYAB antibody is generated from a mouse immunized with a recombination protein from the human region of human CRYAB.
  • Purification:
    Protein G Purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CRYAB
Clonality: Monoclonal
Clone: 1329CT523.140.120
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat