Order Entry
Export
ContactUsLinkComponent
Anti-BCL10 Mouse Monoclonal Antibody [clone: 1185CT13.2.1.2]
Anti-BCL10 Mouse Monoclonal Antibody [clone: 1185CT13.2.1.2]
Catalog # ABGEAM2259B
Supplier:  Abgent
Anti-BCL10 Mouse Monoclonal Antibody [clone: 1185CT13.2.1.2]
Catalog # ABGEAM2259B
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    B-cell CLL/lymphoma 10
  • Antigen symbol:
    BCL10
  • Clonality:
    Monoclonal
  • Clone:
    1185CT13.2.1.2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    8915
  • Antigen synonyms:
    c-E10|IMD37|mE10|CIPER|CLAP|CARMEN
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    26.252 kDa
  • Sequence:
    MEPTAPSLTEEDLTEVKKDALENLRVYLCEKIIAERHFDHLRAKKILSREDTEEISCRTSSRKRAGKLLDYLQENPKGLDTLVESIRREKTQNFLIQKITDEVLKLRNIKLEHLKGLKCSSCEPFPDGATNNLSRSNSDESNF
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.56
  • Shipping temperature:
    4 °C
  • Immunogen:
    This BCL10 antibody is generated from a mouse immunized with a KLH conjugated synthetic peptide between 1-143 amino acids from the human region of human BCL10.
  • Purification:
    Protein G Purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: BCL10
Clonality: Monoclonal
Clone: 1185CT13.2.1.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human